Learn More
5-Methoxy-2-formylphenylboronic acid, TRC
Product Code. 30479607
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30479607 Supplier Toronto Research Chemicals Supplier No. TRCM2634281G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methoxy-2-formylphenylboronic acid
CAS 40138-18-9
Molecular Formula C8H9BO4
Quantity 1 g
Synonym B-(2-Formyl-5-methoxyphenyl)boronic Acid, 2-Formyl-5-methoxyphenylboronic Acid, 5-Methoxy-2-formylphenylboronic Acid
InChI Formula InChI=1S/C8H9BO4/c1-13-7-3-2-6(5-10)8(4-7)9(11)12/h2-5,11-12H,1H3
SMILES COc1ccc(C=O)c(c1)B(O)O
IUPAC Name (2-formyl-5-methoxyphenyl)boronic acid
Molecular Weight (g/mol) 179.97
Formula Weight 180.0594
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.