Learn More
5-Methoxysalicylic acid, 98%
Product Code. 10742011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10742011 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10742011 Supplier Thermo Scientific Acros Supplier No. 163870100
Request Bulk or Custom Format
CAS: 2612-02-4 | C8H8O4 | 168.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 2612-02-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 142°C to 146°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H8O4
Linear Formula CH3OC6H3(OH)CO2H
MDL Number MFCD00002459
Quantity 10 g
Beilstein 10, I, 181
Synonym 5-methoxysalicylic acid, benzoic acid, 2-hydroxy-5-methoxy, 5-methoxy-2-hydroxybenzoic acid, 6-hydroxy-m-anisic acid, m-anisic acid, 6-hydroxy, 2-hydroxy-5-methoxybenzadelhyde, acid5-methoxysalicylic, 2-hydroxy-5-methoxybenzoic, 5-o-methyl gentisic acid, 2-hydroxy-5-methoxy-benzoic acid
Solubility Information Solubility in water: 1.7g/L (10 c)
InChI Key IZZIWIAOVZOBLF-UHFFFAOYSA-N
Molecular Weight (g/mol) 168.15
PubChem CID 75787
Formula Weight 168.15
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 5-Methoxysalicylic acid, 98%