missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1,3,4-thiadiazol-2-thiol, TRC

Product Code. 17910545
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17910545 1 g 1g
30137871 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17910545 Supplier Toronto Research Chemicals Supplier No. TRCM3303051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29490-19-5
Formula Weight 131.9816
InChI Formula InChI=1S/C3H4N2S2/c1-2-4-5-3(6)7-2/h1H3,(H,5,6)
IUPAC Name 5-methyl-1,3,4-thiadiazole-2-thiol
Molecular Formula C3 H4 N2 S2
Molecular Weight (g/mol) 132.21
SMILES Cc1nnc(S)s1
Synonym 5-Methyl-1,3,4-thiadiazol-2-thiol, Cefazolin Sodium Imp. E (EP), MMTD
Chemical Name or Material 5-Methyl-1,3,4-thiadiazol-2-thiol (MMTD)

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.