missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2-furanmethanol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 5-Methyl-2-furanmethanol |
| CAS | 3857-25-8 |
| Molecular Formula | C6 H8 O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H8O2/c1-5-2-3-6(4-7)8-5/h2-3,7H,4H2,1H3 |
| SMILES | Cc1oc(CO)cc1 |
| IUPAC Name | (5-methylfuran-2-yl)methanol |
| Molecular Weight (g/mol) | 112.13 |
| Formula Weight | 112.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction