missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-methylcytosine Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68301-1-IG-150UL
This item is not returnable.
View return policy
Description
The 5-methylcytosine antibody from Proteintech is a mouse monoclonal antibody generated with compound of chemical compound 5-methylcytosine. This antibody recognizes 5mc, chemical compound, m5c antigen. The 5-methylcytosine antibody has been validated for the following applications: IHC, Dot Blot, ELISA analysis.Specifications
| 5-methylcytosine | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| Chemical | |
| Monoclonal Antibody | |
| IgG2b |
| Immunohistochemistry, Dot Blot, ELISA | |
| 2C9G9 | |
| Unconjugated | |
| Compound | |
| 150 μL | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction