Learn More
5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester, 50-70%, see CofA for e, Thermo Scientific™
Product Code. 11916011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11916011 5 g 5g
11906011 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11916011 Supplier Thermo Scientific Acros Supplier No. 439950050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 872054-54-1
Molecular Formula C16H19BN2O2
Molecular Weight (g/mol) 282.15
InChI Key AVMWJACSYAQNDG-UHFFFAOYSA-N
Synonym 5-methylpyridine-2-boronic acid n-phenyldiethanolamine ester, 2-5-methylpyridin-2-yl-6-phenyl-1,3,6,2-dioxazaborocane, 5-methyl-2-pyridineboronic acid n-phenyldiethanolamine ester, 2-5-methyl-pyridin-2-yl-6-phenyl-1,3,6,2 dioxazaborolane, 2-5-methyl-2-pyridinyl-6-phenyl-1,3,6,2-dioxazaborocane, 2-5-methyl-2-pyridinyl-6-phenyl-5,6,7,8-tetrahydro-4h-1,3,6,2-dioxazaborocine
PubChem CID 16217859
IUPAC Name 2-(5-methylpyridin-2-yl)-6-phenyl-1,3,6,2-dioxazaborocane
SMILES B1(OCCN(CCO1)C2=CC=CC=C2)C3=NC=C(C=C3)C
Specifications
Melting Point >300.0°C
Quantity 5 g
Formula Weight 282.15
Percent Purity 50-70%,see CofA for e
Chemical Name or Material 5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester
compliance-icons compliance-icons
Product Identifier
  • 5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.