missing translation for 'onlineSavingsMsg'
Learn More

5-Methylsulfanyl-2-nitroaniline, TRC

Product Code. 30475389
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475389 25 mg 25mg
30478913 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475389 Supplier Toronto Research Chemicals Supplier No. TRCM33196525MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31431-10-4
Molecular Formula C7H8N2O2S
Molecular Weight (g/mol) 184.22
Synonym 5-Methylthio-2-nitroaniline, 5-(Methylthio)-2-nitroaniline
IUPAC Name 5-methylsulfanyl-2-nitroaniline
SMILES CSc1ccc(c(N)c1)[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C7H8N2O2S/c1-12-5-2-3-7(9(10)11)6(8)4-5/h2-4H,8H2,1H3
Formula Weight 184.0306
Chemical Name or Material 5-Methylsulfanyl-2-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.