Learn More
5-(N-Carbamoylamino)imino-4-oxopentanenitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(N-Carbamoylamino)imino-4-oxopentanenitrile |
| CAS | 87015-72-3 |
| Molecular Formula | C6 H8 N4 O2 |
| Quantity | 25 mg |
| Synonym | 2-(4-Cyano-2-oxobutylidene)hydrazine-1-carboxamide |
| InChI Formula | InChI=1S/C6H8N4O2/c7-3-1-2-5(11)4-9-10-6(8)12/h4H,1-2H2,(H3,8,10,12)/b9-4+ |
| SMILES | NC(=O)N\N=C\C(=O)CCC#N |
| IUPAC Name | [(E)-(4-cyano-2-oxo-butylidene)amino]urea |
| Molecular Weight (g/mol) | 168.153 |
| Formula Weight | 168.0647 |