missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Nucleotidase/CD73 Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89705CL1
This item is not returnable.
View return policy
Description
5'-Nucleotidase/CD73 Monoclonal antibody specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| 5'-Nucleotidase/CD73 | |
| Monoclonal | |
| CoraFluor 1 | |
| 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human 5'-Nucleotidase/CD73 (Accession#: NP_002517.1; Met1-Lys547). | |
| 0.1 mL | |
| Lipid and Metabolism, Myeloid derived Suppressor Cell | |
| 4907 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction