missing translation for 'onlineSavingsMsg'
Learn More

5'-Nucleotidase/CD73 Antibody (1E9) [Alexa Fluor™ 594] - Chimeric, Novus Biologicals™

Product Code. 30532909 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532909 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30532909 Supplier Novus Biologicals Supplier No. NBP345121AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

5'-Nucleotidase/CD73 Monoclonal antibody specifically detects 5'-Nucleotidase/CD73 in Human,Mouse (Negative) samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Inhibition Activity,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5'-Nucleotidase/CD73
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 1E9
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Host Species Rabbit
Immunogen Highly purified human placental 5'-Nucleotidase/CD73 (Uniprot#: P21589)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 4907
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.