missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Nucleotidase/CD73 Antibody (45M4F9), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25243JF646
This item is not returnable.
View return policy
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| 5′-Nucleotidase/CD73 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NT5E | |
| Within the range of amino acids 169-324 of human CD73 were used as the immunogen for the antibody. | |
| 0.1 mL | |
| Primary | |
| This product can detect Isoform 1 and Isoform of Human CD73. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 45M4F9 | |
| Western Blot, Immunohistochemistry-Paraffin | |
| 5′ nucleotidase (CD73), 5′-NT, 5′-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5′-nucleotidase, eN, eNT, NT, NT55′-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
| Mouse | |
| Protein G purified | |
| Lipid and Metabolism, Myeloid derived Suppressor Cell | |
| 4907.0 | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction