missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Nucleotidase/CD73 Antibody (AD2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48480B
This item is not returnable.
View return policy
Description
5'-Nucleotidase/CD73 Monoclonal antibody specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Cell Surface)
Specifications
| 5'-Nucleotidase/CD73 | |
| Monoclonal | |
| Biotin | |
| 5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
| Human CD73 [UniProt# P21589] | |
| 0.1 mL | |
| Lipid and Metabolism, Myeloid derived Suppressor Cell | |
| 4907 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry (Cell Surface) | |
| AD2 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction