Learn More
Novus Biologicals™ 5'-Nucleotidase/CD73 Antibody (mupadolimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28558-1mg
 View more versions of this product
Product Code. 30045316
  • € 920.00 / 1mg
Description
5'-Nucleotidase/CD73 Monoclonal antibody specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5'-Nucleotidase/CD73
Monoclonal
Unconjugated
P21589
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
mupadolimab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
NT5E/ CD73
1 mg
Lipid and Metabolism, Myeloid derived Suppressor Cell
4907