missing translation for 'onlineSavingsMsg'
Learn More

5-Oxo Rosuvastatin, TRC

Product Code. 17902405
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17902405 5 mg 5mg
17942405 50 mg 50mg
17904255 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17902405 Supplier Toronto Research Chemicals Supplier No. TRCO8620005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1422619-13-3
Molecular Formula C22 H26 F N3 O6 S
Molecular Weight (g/mol) 479.52
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3-hydroxy-5-oxo-, (3R,6E)-, 5-Oxorosuvastatin, Rosuvastatin Calcium Imp C (EP)
IUPAC Name (E,3R)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3-hydroxy-5-oxohept-6-enoic acid
SMILES CC(C)c1nc(nc(c2ccc(F)cc2)c1\C=C\C(=O)C[C@@H](O)CC(=O)O)N(C)S(=O)(=O)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H26FN3O6S/c1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32/h5-10,13,17,28H,11-12H2,1-4H3,(H,29,30)/b10-9+/t17-/m1/s1
Formula Weight 479.1526
Percent Purity >95%
Chemical Name or Material 5-Oxorosuvastatin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.