missing translation for 'onlineSavingsMsg'
Learn More

5-Oxorosuvastatin Methyl Ester, TRC

Product Code. 17983535
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17983535 1 g 1g
17905045 50 mg 50mg
17923355 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983535 Supplier Toronto Research Chemicals Supplier No. TRCO8620051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147118-39-6
Molecular Formula C23 H28 F N3 O6 S
Molecular Weight (g/mol) 493.55
Synonym 5-Oxorosuvastatin Methyl Ester, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3-hydroxy-5-oxo-, methyl ester, (3R,6E)-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3-hydroxy-5-oxo-, methyl ester, [R-(E)]-, Methyl (+)-(3R)-7-[4-(4-fluorophenyl)-6-isopropyl-2-(N-methyl-N-Methansulphonylamino)pyrimidin-5-yl]-3-hydroxy-5-oxo-6(E)-heptenoate, Methyl (3R,6E)-7-{4-(4-fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl}-3-hydroxy-5-oxo-6-heptenoate
IUPAC Name methyl (E,3R)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3-hydroxy-5-oxohept-6-enoate
SMILES COC(=O)C[C@H](O)CC(=O)\C=C\c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C23H28FN3O6S/c1-14(2)21-19(11-10-17(28)12-18(29)13-20(30)33-4)22(15-6-8-16(24)9-7-15)26-23(25-21)27(3)34(5,31)32/h6-11,14,18,29H,12-13H2,1-5H3/b11-10+/t18-/m1/s1
Formula Weight 493.1683
Chemical Name or Material 5-Oxorosuvastatin Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.