Learn More
5-Pyrimidinemethanamine, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Pyrimidinemethanamine |
| CAS | 25198-95-2 |
| Molecular Formula | C5 H7 N3 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C5H7N3/c6-1-5-2-7-4-8-3-5/h2-4H,1,6H2 |
| SMILES | NCc1cncnc1 |
| IUPAC Name | pyrimidin-5-ylmethanamine |
| Molecular Weight (g/mol) | 109.13 |
| Formula Weight | 109.064 |