Learn More
5-Sulphamoylsalicylic Acid, Mikromol™
Product Code. 30052233
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052233 100 mg 100mg
30051843 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30052233 Supplier Mikromol Supplier No. MM0051.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Sulphamoylsalicylic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Sulphamoylsalicylic Acid
CAS 5378-41-6
Analyte or Component Names 5-Sulphamoylsalicylic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H7 N O5 S
Certifications/Compliance ISO 9001
Synonym 5-Sulphamoylsalicylic Acid
InChI Formula InChI=1S/C7H7NO5S/c8-14(12,13)4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,(H,10,11)(H2,8,12,13)
SMILES NS(=O)(=O)c1ccc(O)c(c1)C(=O)O
IUPAC Name 2-hydroxy-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 217.2
Quantity 100 mg
Formula Weight 217.0045 g/mol