missing translation for 'onlineSavingsMsg'
Learn More

5-Thiazolecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7077490
83.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14527-41-4
C4H3NO2S
129.13
MFCD03428539
YZVFSQQHQPPKNX-UHFFFAOYSA-N
thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, thiazole-5-carboxylicacid, pubchem14533, acmc-209cuh, 5-thiazole carboxylic acid, thiazole 5-carboxylic acid, ksc177g9t
84494
1,3-thiazole-5-carboxylic acid
OC(=O)C1=CN=CS1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14527-41-4
C4H3NO2S
129.13
MFCD03428539
YZVFSQQHQPPKNX-UHFFFAOYSA-N
thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, thiazole-5-carboxylicacid, pubchem14533, acmc-209cuh, 5-thiazole carboxylic acid, thiazole 5-carboxylic acid, ksc177g9t
84494
1,3-thiazole-5-carboxylic acid
OC(=O)C1=CN=CS1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 208.0°C to 210.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.14
Percent Purity 98%
Chemical Name or Material 5-Thiazolecarboxylic acid
compliance-icons
Product Identifier
  • 5-Thiazolecarboxylic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'