missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BETP

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective positive allosteric modulator and partial agonist of the glucagon-like peptide 1 (GLP-1) receptor. Increases binding affinity of oxyntomodulin for the GLP-1 receptor. Potentiates oxyntomodulin-mediated GLP-1 receptor signaling in vitro and insulin secretion in vivo. Has no effect on GLP-2, GIP, PTH or glucagon receptors.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 406.42 |
| Molecular Weight (g/mol) | 406.423 |
| Chemical Name or Material | BETP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction