Learn More
SI Analytics™ 50 mL Exchangeable Unit
Product Code. 13486979 Shop All SI Analytics Products
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Each
Product Code. 13486979
Brand: SI Analytics™ 285220350
Only null left
Add to basket
Add to basket
SI Analytics™ 50 mL Exchangeable Unit is an accessory for TitroLine™ and Titronic™ models.
  • With integrated chip for reagent data and brown glass bottle for titrant
  • Includes GL 45 and S 40-bottle adapter
TRUSTED_SUSTAINABILITY
Specifications
Type WA 50
Includes Integrated chip, glass bottle, bottle adapter, drip tubes, and titration itp
Volume (Metric) 50 mL