Learn More
Labcon™ PerformR™ 50 mL Polypropylene Centrifuge Tubes
GREENER_CHOICE
Product Code. 30533294
Click to view available options
Closure Type:
Attached Flat Screw Cap
Plug
Plug Style Screw Cap
Sterility:
Non-sterile
Sterile
Quantity:
25 Tubes/Plastic Rack
25 Tubes/Reusable Rack, 12 Racks/Cs.
50 Tubes/Bag, 10 Bags/Cs.
500 Tubes and Caps in Separate Bags
500 Tubes/Bag, 500 Caps in Separate Bag, 500 Tubes and Caps/Cs.
Unit Size:
Pack of 25
Pack of 300
Pack of 50
Pack of 500
Product Code. 30533294
Brand: Labcon™ 31873450069
Edge
Centrifuge tubes are designed for everyday centrifugation and storage needs. Available with either a flat cap or a plug style cap.
  • Centrifuging to 12,500 x G
  • Freezable to -80°C
  • Tubes are autoclavable
  • Made in USA
Specifications
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Closure Material Polyethylene
Closure Color Purple
Color Natural
Capacity (Metric) 50 mL
Disposable Single-use
Material Polypropylene
For Use With Centrifuges, Rotators, Shakers
No. per Pack 50/Pk.
Packaging 50 Tubes/Bag, 10 Bags/Cs.
Quantity 50 Tubes/Bag, 10 Bags/Cs.
Sterility Sterile
Product Line PerformR
RNase Free Yes
Dnase Free Yes
Autoclavable Autoclavable
Shape Cylindrical
Length (Metric) 115 mm
Clean Claims ATP, RNase/DNase and Endotoxin-free
ACT Label Yes
Bottom Shape Conical
Closure Type Plug
Closure Included Yes
Writing Area Yes
Height (Metric) 115 mm
Diameter (Metric) 29 mm
Graduated Yes
Graduations 5 to 45 mL in 2.5 mL Increments
Product Type Centrifuge Tube
Max. RCF 12,500 x g