missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human MUC-1 Fc Chimera Protein

Product Code. 18718196
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18718196

Brand: R&D Systems™ 10332MU050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human MUC-1 Fc Chimera is immobilized at 1 μg/mL (100 μL/well), Recombinant Human Siglec-9 Fc Chimera binds with an ED50 of 0.2 to 1.4 μg/mL
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15941.3
For Use With (Application) Bioactivity
Gene ID (Entrez) 4582
Molecular Weight (g/mol) M.W. (Observed): 157 - 192 kDa, under reducing conditions.
Name Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEM, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human MUC-1 protein Human MUC-1 (Ser24-Ser380) (C-terminus) Accession # P15941.3 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 400 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <1.0 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.