Learn More
Hamilton™ 1 mL 1001 Series Gas Tight Syringe Luer Tip (LT)
Product Code. 17824422
Change view
Click to view available options
:
Needle Gauge: 22 G, Point Style: 3, Connection Type: RN
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17824422 Needle Gauge: 22 G, Point Style: 3, Connection Type: RN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17824422 Supplier Hamilton™ Supplier No. CAL81301
Only null left
Add to basket
Add to basket
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
  • This series of syringes are ideal for dispensing volumes up to 500 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass
Description Luer Tip (LT)
Diameter (English) Inner 0.181 in.
Diameter (English) Outer 0.355 in.
Diameter (Metric) Inner 3.26 mm
Diameter (Metric) Outer 7.75 mm
Product Type Gas Tight Syringe
Max. Temperature (English) 176°F
Max. Temperature (Metric) 80°C
Min. Temperature (English) 50°F
Model 1750
Min. Temperature (Metric) 10°C
Quantity 1
Volume (Metric) 1 mL
Needle Point Style Kel-F Hub
Connection Type Removable