missing translation for 'onlineSavingsMsg'
Learn More

Butralin, TRC

Product Code. 30335270
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30335270

Brand: Toronto Research Chemicals TRCB694350500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butralin
CAS 33629-47-9
Purity Grade Notes HPLC
Molecular Formula C14 H21 N3 O4
Quantity 500 mg
Synonym Aniline, N-sec-butyl-4-tert-butyl-2,6-dinitro- (8CI), 4-(1,1-Dimethylethyl)-N-(1-methylpropyl)-2,6-dinitrobenzenamine, 4-tert-Butyl-N-mono-sec-butyl-2,6-dinitroaniline, 4-tert-Butyl-N-sec-butyl-2,6-dinitroaniline, 70-314B, 72-A34, A 820, A 820 (herbicide), Amchem 70-25, Amex, Amex 820, Butalin, Butralin, Dibutalin, N-sec-Butyl-4-tert-butyl-2,6-dinitroaniline
InChI Formula InChI=1S/C14H21N3O4/c1-6-9(2)15-13-11(16(18)19)7-10(14(3,4)5)8-12(13)17(20)21/h7-9,15H,6H2,1-5H3
SMILES CCC(C)Nc1c(cc(cc1[N+](=O)[O-])C(C)(C)C)[N+](=O)[O-]
IUPAC Name N-butan-2-yl-4-tert-butyl-2,6-dinitroaniline
Molecular Weight (g/mol) 295.33
Formula Weight 295.1532
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.