missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylhexylphthalate (Bis-(2-ethylhexyl) Phthalate), Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Diethylhexylphthalate (Bis-(2-ethylhexyl) Phthalate)
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-2-ethylhexyl ester |
| CAS | 117-81-7 |
| Analyte or Component Names | Phthalic acid, bis-2-ethylhexyl ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H38 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Diethylhexylphthalate, Bis-(2-ethylhexyl) Phthalate, Plastic additive 01, DEHP |
| InChI Formula | InChI=1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3 |
| SMILES | CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC |
| IUPAC Name | bis(2-ethylhexyl) benzene-1,2-dicarboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction