Learn More
Testosterone Laurate, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Testosterone Laurate |
| CAS | 59232-78-9 |
| Molecular Formula | C31 H50 O3 |
| Quantity | 500 mg |
| Synonym | (17β)-17-[(1-oxododecyl)oxy]-Androst-4-en-3-one, Testosterone','laurate (6CI), Testosterone Laurate, 3-Oxoandrost-4-en-17β-yl dodecanoate |
| InChI Formula | InChI=1S/C31H50O3/c1-4-5-6-7-8-9-10-11-12-13-29(33)34-28-17-16-26-25-15-14-23-22-24(32)18-20-30(23,2)27(25)19-21-31(26,28)3/h22,25-28H,4-21H2,1-3H3/t25-,26-,27-,28-,30-,31-/m0/s1 |
| SMILES | CCCCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] dodecanoate |
| Molecular Weight (g/mol) | 470.7269 |
| Formula Weight | 470.376 |