Learn More
Polysulfide Oil Calibration Sample: Sulfur - S @ 100μg/g in Light Mineral Oil, VHG™
Product Code. 17982416
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
Product Code. 17982416
Brand: VHG Labs VHGPS20M100500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Polysulfide Oil Calibration Sample: Sulfur - S @ 100μg/g in Light Mineral Oil
TRUSTED_SUSTAINABILITY
Specifications
Methods XRF
Shipping Condition Room Temperature
Solvent or Matrix 20 cSt Mineral Oil
Quantity 500 mL