Learn More
Thermo Scientific 500ML SOLUTION TUNE B iCAPq
Brand:  Thermo Scientific 1323770
500ML SOLUTION TUNE B iCAPq
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 13502810
  • POA / 500mL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • iCAP Q/RQ TUNE solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, hydrochloric acid