Learn More
Sulfate Standard: SO4(-2) @ 100 μg/mL in H2O, VHG™
Product Code. 17907934
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 17907934
Brand: VHG Labs VHGISO4100500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfate Standard: SO4(-2) @ 100 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfate
CAS 14808-79-8
Methods Ion Chromatography/Wet Chemistry
Analyte or Component Names Sulfate
Shipping Condition Room Temperature
Molecular Formula (S O4)2-
Certifications/Compliance ISO 17034
Synonym SO42-
SMILES [O-]S(=O)(=O)[O-]
IUPAC Name Sulfate
Solvent or Matrix Water
Molecular Weight (g/mol) 96.06
Quantity 500 mL
Formula Weight 96.06 g/mol