Learn More
Viscosity Standard: 73 cSt at 40°C, and 9 cSt at 100°C, VHG™
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Viscosity Standard: 73 cSt at 40°C, and 9 cSt at 100°C
Specifications
Specifications
| Chemical Name or Material | Viscosity |
| Methods | Viscometer |
| Analyte or Component Names | Viscosity |
| Shipping Condition | Room Temperature |
| Concentration | 73 cSt |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hydrocarbons |
| Quantity | 500 mL |