Learn More
Spectrum™ Labs Spectra/Por™ 5 12-14 kD MWCO Standard RC Dry Dialysis Kits
Description
Includes
5 m of Spectra/Por 5 dialysis tubing and 2 closures
- For reinforced for weight applications, or evaluating dialysis tubing for dialysis application
- Packaged dry without glycerin (no soaking required)
Specifications
Specifications
| Product Type | Dialysis Tubing |
| MWCO | 12 to 14 kDa |
| Format | Tubing |
| Diameter (Metric) | 48 mm |
| Length (Metric) | 18 mL/cm vol./length |
| Quantity | 5 m Tubing/Kit |