missing translation for 'onlineSavingsMsg'
Learn More

Bumetrizole, TRC

Product Code. 30343633
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30343633

Brand: Toronto Research Chemicals TRCB68968050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bumetrizole
CAS 3896-11-5
Molecular Formula C17 H18 Cl N3 O
Quantity 50 g
Synonym p-Cresol, 2-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)- (7CI,8CI), 2-(5-Chloro-2H-benzotriazol-2-yl)-6-(1,1-dimethylethyl)-4-methylphenol, 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chloro-2H-benzotriazole, 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chlorobenzotriazole, 2-(2'-Hydroxy-3'-tert-butyl-5'-methylphenyl)-5-chlorobenzotriazole, 2-(2'-Hydroxy-3'-tert-butyl-5'-methylphenyl)-5-chlorobenztriazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chloro-2H-benzo-v-triazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chloro-2H-benzotriazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chlorobenzotriazole, 2-(3-tert-Butyl-5-methyl-2-hydroxyphenyl)-5-chlorobenzotriazole, 2-(3'-tert-Butyl-2'-hydroxy-5'-methylphenyl)-5-chlorobenzotriazole, 2-(3'-tert-Butyl-5'-methyl-2'-hydroxyphenyl)-5-chlorobenzotriazole, 2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol, 2-(5-Chloro-2H-benzotriazol-2-yl)-6-tert-butyl-4-methylphenol, 2-[5-Chloro-2H-benzotriazol-2-yl]-4-methyl-6-(tert-butyl)phenol, 2-tert-Butyl-6-(5-chloro-2-benzotriazolyl)-4-methylphenol, 2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol, 5-Chloro-2-(2-hydroxy-3-tert-butyl-5-methylphenyl)-2H-benzotriazole, 5-Chloro-2-(3-tert-butyl-2-hydroxy-5-methylphenyl)benzotriazole, A-Light, ADK Stab LA 36, ADK Stab LA 36RG, Benazol PBKh, Bumetrizole, CBT, Cibatex LF, Eversorb 73, J 395, J 395 (light stabilizer), JC 30S, JF 79, Kemisorb 73, LA 36, Light Stabilizer 326, Lowilite 26, Mark LA 36, Seesorb 703, Songsorb 3260, Sumisorb 300, T 326, TNV 326, Tin 326, Tinogard AS, Tinuvin 326, Tomisorb 600, UV 326, Uvinul 3026, VS 550, Viosorb 550
InChI Formula InChI=1S/C17H18ClN3O/c1-10-7-12(17(2,3)4)16(22)15(8-10)21-19-13-6-5-11(18)9-14(13)20-21/h5-9,22H,1-4H3
SMILES Cc1cc(c(O)c(c1)C(C)(C)C)n2nc3ccc(Cl)cc3n2
IUPAC Name 2-tert-butyl-6-(5-chlorobenzotriazol-2-yl)-4-methylphenol
Molecular Weight (g/mol) 315.797
Formula Weight 315.1138
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.