missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Methylpropyl)-4-[(3RS)-3-[4-(2-methylpropyl)phenyl]butyl]benzene, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
1-(2-Methylpropyl)-4-[(3RS)-3-[4-(2-methylpropyl)phenyl]butyl]benzene
Specifications
Specifications
| Chemical Name or Material | 1-(2-Methylpropyl)-4-[(3RS)-3-[4-(2-methylpropyl)phenyl]butyl]benzene |
| CAS | 2143535-26-4 |
| Analyte or Component Names | 1-(2-Methylpropyl)-4-[(3RS)-3-[4-(2-methylpropyl)phenyl]butyl]benzene |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H34 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-(2-Methylpropyl)-4-[(3RS)-3-[4-(2-methylpropyl)phenyl]butyl]benzene, Ibuprofen Imp. I (EP), Ibuprofen Impurity I |
| InChI Formula | InChI=1S/C24H34/c1-18(2)16-22-10-8-21(9-11-22)7-6-20(5)24-14-12-23(13-15-24)17-19(3)4/h8-15,18-20H,6-7,16-17H2,1-5H3 |
| SMILES | CC(C)Cc1ccc(CCC(C)c2ccc(CC(C)C)cc2)cc1 |
| IUPAC Name | 1-(2-methylpropyl)-4-[4-[4-(2-methylpropyl)phenyl]butan-2-yl]benzene |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction