missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1H-1,2,4-Triazol-1-yl)acetic Acid-D4, TRC
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Specifications
Specifications
| Chemical Name or Material | 2-(1H-1,2,4-Triazol-1-yl)acetic Acid-D4 |
| Molecular Formula | C4 D4 H N3 O2 |
| Quantity | 50 mg |
| Synonym | 2-(1H-1,2,4-Triazol-1-yl-D2)acetic-2,2-D2 Acid |
| InChI Formula | InChI=1S/C4H5N3O2/c8-4(9)1-7-3-5-2-6-7/h2-3H,1H2,(H,8,9)/i1D2,2D,3D |
| SMILES | [2H]c1nc([2H])n(n1)C([2H])([2H])C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-(3,5-dideuterio-1,2,4-triazol-1-yl)acetic acid |
| Molecular Weight (g/mol) | 131.126 |
| Formula Weight | 131.063 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction