missing translation for 'onlineSavingsMsg'
Learn More

2,2'-(Piperazine-1,4-diyl)dipyrimidine, Mikromol™

Product Code. 30051095
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30051095

Brand: Mikromol MM0464.12

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,2'-(Piperazine-1,4-diyl)dipyrimidine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2'-(Piperazine-1,4-diyl)dipyrimidine
CAS 84746-24-7
Analyte or Component Names 2,2'-(Piperazine-1,4-diyl)dipyrimidine
Shipping Condition Room Temperature
Molecular Formula C12 H14 N6
Certifications/Compliance ISO 17025
Synonym 2,2'-(Piperazine-1,4-diyl)dipyrimidine, Buspirone Hydrochloride Imp. G (EP), Buspirone Related Compound G, Buspirone Hydrochloride Impurity G, Buspirone Impurity G
InChI Formula InChI=1S/C12H14N6/c1-3-13-11(14-4-1)17-7-9-18(10-8-17)12-15-5-2-6-16-12/h1-6H,7-10H2
SMILES C1CN(CCN1c2ncccn2)c3ncccn3
IUPAC Name 2-(4-pyrimidin-2-ylpiperazin-1-yl)pyrimidine
Molecular Weight (g/mol) 242.28
Quantity 50 mg
Formula Weight 242.128 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.