missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl]methanol, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
[2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl]methanol
Specifications
Specifications
| Chemical Name or Material | (2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl)methanol |
| CAS | 114799-13-2 |
| Analyte or Component Names | (2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl)methanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H23 Cl N6 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Losartan Potassium Imp. C (EP), Losartan Imp. C (EP), [2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl]methanol, Losartan Potassium Impurity C, Losartan Impurity C |
| InChI Formula | InChI=1S/C22H23ClN6O/c1-2-3-8-20-24-19(14-30)21(23)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22/h4-7,9-12,30H,2-3,8,13-14H2,1H3,(H,25,26,27,28) |
| SMILES | CCCCc1nc(CO)c(Cl)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4 |
| IUPAC Name | [2-butyl-5-chloro-1-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction