missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-(prop-2-enyl)-10,11-dihydro-5H-dibenzo[b,f]azepine, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
3-Chloro-5-(prop-2-enyl)-10,11-dihydro-5H-dibenzo[b,f]azepine
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-5-(prop-2-enyl)-10,11-dihydro-5H-dibenzo[b,f]azepine |
| CAS | 1425793-87-8 |
| Analyte or Component Names | 3-Chloro-5-(prop-2-enyl)-10,11-dihydro-5H-dibenzo[b,f]azepine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H16 Cl N |
| Certifications/Compliance | ISO 9001 |
| Synonym | Clomipramine Hydrochloride Imp. G (EP), Clomipramine Imp. G (EP), 3-Chloro-5-(prop-2-enyl)-10,11-dihydro-5H-dibenzo[b,f]azepine, Clomipramine Hydrochloride Impurity G, Clomipramine Impurity G, N-Allyl-3-chloro-10,11-dihydro-5H-dibenzo[b,f]azepine |
| InChI Formula | InChI=1S/C9H15NO/c11-8-6-9(7-10-8)4-2-1-3-5-9/h1-7H2,(H,10,11) |
| SMILES | Clc1ccc2CCc3ccccc3N(CC=C)c2c1 |
| IUPAC Name | 2-chloro-11-prop-2-enyl-5,6-dihydrobenzo[b][1]benzazepine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction