missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylindolin-2-one (4-Ethenyl-1,3-dihydro-2H-indol-2-one), Mikromol™

Product Code. 17932496
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 17932496

Brand: Mikromol MM0331.11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Vinylindolin-2-one (4-Ethenyl-1,3-dihydro-2H-indol-2-one)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Ethenyl-1,3-dihydro-2H-indol-2-one
CAS 120427-93-2
Analyte or Component Names 4-Ethenyl-1,3-dihydro-2H-indol-2-one
Shipping Condition Ice pack (-20°C)
Molecular Formula C10 H9 N O
Certifications/Compliance ISO 9001
Synonym 4-Ethenyl-1,3-dihydro-2H-indol-2-one
InChI Formula InChI=1S/C10H9NO/c1-2-7-4-3-5-9-8(7)6-10(12)11-9/h2-5H,1,6H2,(H,11,12)
SMILES C=Cc1cccc2NC(=O)Cc12
IUPAC Name 4-ethenyl-1,3-dihydroindol-2-one
Molecular Weight (g/mol) 159.18
Quantity 50 mg
Formula Weight 159.0684 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.