missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
7-Bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one
Specifications
Specifications
| Chemical Name or Material | 7-Bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one |
| CAS | 868350-97-4 |
| Analyte or Component Names | 7-Bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H12 Br N3 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 7-Bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Bromazepam Imp. C (EP), Bromazepam Impurity C |
| InChI Formula | InChI=1S/C15H12BrN3O/c1-9-3-2-4-13(18-9)15-11-7-10(16)5-6-12(11)19-14(20)8-17-15/h2-7H,8H2,1H3,(H,19,20) |
| SMILES | Cc1cccc(n1)C2=NCC(=O)Nc3ccc(Br)cc23 |
| IUPAC Name | 7-bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-1,4-benzodiazepin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction