Learn More
Benzo[e]pyrene, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Benzo(e)pyrene |
| CAS | 192-97-2 |
| Molecular Formula | C20 H12 |
| Quantity | 50 mg |
| Synonym | Benzo[e]pyrene, 1,2-Benzopyrene, 1,2-Benzpyrene, 4,5-Benzopyrene, 4,5-Benzpyrene, NSC 89273 |
| InChI Formula | InChI=1S/C20H12/c1-2-8-16-15(7-1)17-9-3-5-13-11-12-14-6-4-10-18(16)20(14)19(13)17/h1-12H |
| SMILES | c1ccc2c(c1)c3cccc4ccc5cccc2c5c34 |
| IUPAC Name | benzo[e]pyrene |
| Molecular Weight (g/mol) | 252.31 |
| Formula Weight | 252.0939 |