missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fmoc-L-azidoalanine, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Fmoc-L-azidoalanine |
| CAS | 684270-46-0 |
| Molecular Formula | C18 H16 N4 O4 |
| Quantity | 50 mg |
| Synonym | 3-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-alanine |
| InChI Formula | InChI=1S/C18H16N4O4/c19-22-20-9-16(17(23)24)21-18(25)26-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15-16H,9-10H2,(H,21,25)(H,23,24)/t16-/m0/s1 |
| SMILES | OC(=O)[C@H](CN=[N+]=[N-])NC(=O)OCC1c2ccccc2c3ccccc13 |
| IUPAC Name | (2S)-3-azido-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid |
| Molecular Weight (g/mol) | 352.34 |
| Formula Weight | 352.1172 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction