missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lactic Acid-13C3 Sodium Salt (20% W/W in H2O), TRC

Product Code. 17967264
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17967264 50 mg 50mg
17968314 10 mg 10mg
17958424 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17967264 Supplier Toronto Research Chemicals Supplier No. TRCL11350850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201595-71-3
Molecular Formula 13C3 H5 O3 . Na
Molecular Weight (g/mol) 115.04
Synonym (2S)-2-Hydroxy-propanoic Acid-13C3 Sodium Salt, (+)-Lactic Acid-13C3 Sodium Salt, (S)-(+)-Lactic Acid-13C3 Sodium Salt, (S)-2-Hydroxypropanoic Acid-13C3 Sodium Salt, (S)-2-Hydroxypropionic Acid-13C3 Sodium Salt, (S)-Lactic Acid-13C3 Sodium Salt, Espiritin-13C3 Sodium Salt, HiPure 90-13C3 Sodium Salt;Paralactic Acid-13C3 Sodium Salt, Sarcolactic Acid-13C3 Sodium Salt, Tisulac-13C3 Sodium Salt, Sodium L-Lactate-13C3
IUPAC Name sodium;(2S)-2-hydroxy(1,2,3-^{13}C_{3})propanoate
SMILES [Na+].[13CH3][13C@H](O)[13C](=O)[O-]

Specifications

Solvent or Matrix Water
Quantity 50 mg
InChI Formula InChI=1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i1+1,2+1,3+1;
Formula Weight 115.0237
Concentration or Composition (by Analyte or Components) L-(+)-Lactic Acid-13C3 Sodium Salt 20 %
Chemical Name or Material L-(+)-Lactic Acid-13C3 Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.