Learn More
Tocris Bioscience™ Pregnenolone sulfate sodium salt
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Pregnenolone sulfate sodium salt is a highly potent neurosteroid with multiple actions. Enhances intracellular Ca2+ upon NMDA receptor-mediated synaptic activity (EC50 ∽ 2 pM) and potentiates NMDA responses. Inhibitor at GABAA receptors (IC50 values are 0.4 to >300 μM, depending on specific subunit conformation). Activates CREB phosphorylation, augments LTP and cognitive behavior in rat memory models. Also activates TRPM3 channels.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | Pregnenolone sulfate sodium salt |
| Molecular Weight (g/mol) | 418.52 |
| Inhibitors | Pregnenolone sulfate sodium salt |