missing translation for 'onlineSavingsMsg'
Learn More

0 wt% Soot in Diesel Engine Oil, VHG™

Product Code. 17928804
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 17928804

Brand: VHG Labs VHGSOOTBLK50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

0 wt% Soot in Diesel Engine Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nominal Soot Content Range: Blank
Methods FTIR/IR
Analyte or Component Names Nominal Soot Content Range: Blank
Shipping Condition Room Temperature
IUPAC Name Nominal Soot Content Range: Blank
Solvent or Matrix 15W40 Motor Oil
Quantity 50 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.