missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ 50TS Microplate Washer

For biomagnetic separation and vacuum filtration

Brand:  BioTek™ 50TS8V

Additional Details : Weight : 10.00000kg

 View more versions of this product

Product Code. 15741726

  • € 6357.00 / Each


Estimated Shipment
01-12-2020
Add to basket

Description

Description

  • For 96-well washing
  • Automated
  • 12-well manifold
  • 4.3 in. color touchscreen LCD
  • 1 to 10 wash cycles

Warranty

12 months

Specifications

Specifications

1 to 10 wash cycles
96-well washing
Microplate Strip Washer
40.6D x 35.6W x 16.5cm H
4.3 in. color touchscreen LCD
With buffer switching
Automated
8-well manifold
Videos
SDS
Documents