missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-305AF405
This item is not returnable.
View return policy
Description
53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse, Rat, Bat, Bovine, Canine, Naked mole-rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| 53BP1 | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Q12888 | |
| TP53BP1 | |
| The epitope recognized by 53BP1 Antibody maps to a region between residues 1925 and the C-terminus (residue 1972) of human 53BP1 (NP_005648.1). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Tumor Suppressors | |
| 7158 | |
| Human, Mouse, Rat, Bat, Bovine, Canine | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human has been tested in both WB and ICC/IF, mouse has only been tested in ICC/IF. Rat reactivity reported in scientific literature (PMID: 24244353). Feedback on bovine has been negative. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction