missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-904MFV610
This item is not returnable.
View return policy
Description
53BP1 Polyclonal antibody specifically detects 53BP1 in Human, Mouse, Other samples. It is validated for Flow Cytometry
Specifications
| 53BP1 | |
| Polyclonal | |
| Flow Cytometry | |
| 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
| 53BP1 Antibody was made to a synthetic peptide corresponding to a portion of human 53BP1 encoded in exon 11, 12 and 19 [Uniprot: Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Tumor Suppressors | |
| 7158 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Other | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction