missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 53BP1 Polyclonal Antibody, HRP

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522793
This item is not returnable.
View return policy
Description
This antibody reacts with Human samples in Western blot and ICC applications, and with Mouse in ICC applications. Predicted cross-reactivity based on sequence identity: Chimpanzee/Orangutan (99%), Gibbon/Gorilla (98%), Marmoset (95%).
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
Specifications
| 53BP1 | |
| Polyclonal | |
| HRP | |
| TP53BP1 | |
| 53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27223, 7158 | |
| 4°C, store in dark | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot, Proximity Ligation Assay, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| P70399, Q12888 | |
| TP53BP1, Trp53bp1 | |
| Synthetic peptide corresponding to a portion of human 53BP1 encoded in exon 11, 12 and 19. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction