Learn More
5a-Androstane-3b,17b-diol, TRC
Specifications
Specifications
| Chemical Name or Material | 5a-Androstane-3b,17b-diol |
| CAS | 571-20-0 |
| Molecular Formula | C19H32O2 |
| Quantity | 500 mg |
| Synonym | Androstane-3,17-diol, (3β,5α,17β)-, 5α-Androstane-3β,17β-diol (7CI,8CI), (3β,5α,17β)-Androstane-3,17-diol, 3β,17β-Androstanediol, 3β,17β-Dihydroxy-5α-androstane, 3β-Adiol, Maxterone, NSC 50891 |
| InChI Formula | InChI=1S/C19H32O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-17,20-21H,3-11H2,1-2H3/t12-,13-,14-,15-,16-,17-,18-,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CC[C@H]4C[C@@H](O)CC[C@]34C)[C@@H]1CC[C@@H]2O |
| IUPAC Name | (3S,5S,8R,9S,10S,13S,14S,17S)-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 292.4 |
| Formula Weight | 292.4562 |