missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Androstan-3alpha-ol-17-one-16,16-d2, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Androsterone-16,16-D2 |
| CAS | 89685-22-3 |
| Molecular Formula | C19 D2 H28 O2 |
| Quantity | 0.1 g |
| Synonym | Androstan-17-one-16,16-d2, 3-hydroxy-, (3α,5α)- (9 CI), 5α-Androstan-3α-ol-17-one-16,16-d2, Androsterone-d2, Androsterone-16,16-d2, Androsterone-16,16-D2, (3α,5α)-3-Hydroxyandrostan-17-one-16,16-d2 |
| InChI Formula | InChI=1 S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20 H,3-11H2,1-2H3/t12-,13+,14-,15-,16-,18-,19-/m0/s1/i6D2 |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CC[C@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 CC[C@]2(C)C1=O |
| IUPAC Name | (3 R,5 S,8 R,9 S,10 S,13 S,14 S)-16,16-dideuterio-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15-dodecahydro-1 H-cyclopenta[a]phenanthren-17-one |
| Molecular Weight (g/mol) | 292.4526 |
| Formula Weight | 292.2371 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction